Merck
GC Analysis of Volatile Flavor Compound Enantiomers on Astec® CHIRALDEX G-TA
GC Analysis of Volatile Flavor Compound Enantiomers on Astec® CHIRALDEX™ G-TA suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX G-TA Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
Methyl DL-lactate
≥97.0% (GC)
1-Phenylethanol
98%
α-Ionone
natural, ≥86%
(R)-(−)-Carvone
98%
Methyl DL-mandelate
97%
β-Butyrolactone
98%
δ-Octanolactone
AldrichCPR
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven
60 °C, 2 °C/ min to 160 °C (hold 20 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 cm/sec, set at 75 °C
injection
1 μL, split 100:1
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC