GC Analysis of FAMEs in Atlantic Salmon on SLB®-IL60
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL60, 30 m x 0.25 mm I.D., 0.20 μm (29505-U)
oven
170 °C, 1 °C/min to 225 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 1.2 mL/min
injection
1 μL 100:1 split
liner
4 mm I.D., split/splitless type, wool packed single taper FocusLiner™ design
sample
extract of a farm raised Atlantic salmon, the antioxidant butylated hydroxytoluene (BHT) added prior to extraction
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany