GC Analysis of Class 2 Residual Solvents on Equity-1 after SPME using 85 μm Polyacrylate Fiber
Materials
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly polyacrylate (PA)
df 85 μm(PA, for use with manual holder, needle size 24 gaCONDITIONS
sample/matrix
2 mL water + sodium chloride at 25% (pH 11) containing each analyte at 5 ppm, in a 4 mL vial
SPME fiber
85 μm Polyacrylate, 24 ga (57304)
extraction
eated headspace, 60 °C for 5 min
desorption flow
250 °C for 3 min
column
Equity-1, 10 m x 0.20 mm I.D., 1.2 μm (28043-U)
oven
40 °C (0.75 min), 20 °C/min to 200 °C (10 min)
inj. temp.
250 °C
detector
FID
carrier gas
helium, 35 cm/sec @ 40 °
injection
5:1 split
liner
0.75 mm I.D. SPME type
Description
Legal Information
null