GC Analysis of Alcohols and Hydrocarbons on Equity-1
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Equity-1, 30 m × 0.53 mm I.D., 3.0 μm (28076-U)
oven
40 °C (5 min), 8 ºC/min to 225 °C (10 min)
inj. temp.
250 °C
detector
FID, 275 °C
carrier gas
helium, 30 cm/sec @ 40 °C
injection
0.1 μL, split 100:1
liner
split, cup design
sample
32 component mixed solvent sample, equal by weight
Description
Legal Information
null