Merck
  • Cyclooxygenase 2 expression in colorectal cancer with DNA mismatch repair deficiency.
Cyclooxygenase 2 expression in colorectal cancer with DNA mismatch repair deficiency.
Clinical cancer research : an official journal of the American Association for Cancer Research (2006-03-23)
Antoni Castells, Artemio Payá, Cristina Alenda, Francisco Rodríguez-Moranta, Rubén Agrelo, Montserrat Andreu, Virgínia Piñol, Sergi Castellví-Bel, Rodrigo Jover, Xavier Llor, Elisenda Pons, J Ignasi Elizalde, Xavier Bessa, Javier Alcedo, Joan Saló, Enrique Medina, Antonio Naranjo, Manel Esteller, Josep M Piqué
ABSTRACT
Cyclooxygenase 2 (COX-2) overexpression is a frequent but not universal event in colorectal cancer. It has been suggested that COX-2 protein expression is reduced in colorectal cancer with a defective mismatch repair (MMR) system, a phenomenon commonly associated with hereditary nonpolyposis colorectal cancer (HNPCC) but also present in up to 15% of sporadic tumors. To assess COX-2 expression in a large series of fully characterized colorectal cancer patients with respect to the MMR system and to dissect the mechanisms responsible for altered COX-2 expression in this setting. MMR-deficient colorectal cancer were identified in a nationwide, prospective, multicenter study (EPICOLON project). Control MMR-proficient colorectal cancer patients were randomly selected. COX-2 expression was evaluated by immunohistochemistry. Personal and familial characteristics, as well as MSH2/MLH1 expression and germ line mutations, were evaluated. One hundred fifty-three patients, 46 with MMR deficiency and 107 with MMR proficiency, were included in the analysis. Overall, tumor COX-2 overexpression was observed in 107 patients (70%). COX-2 overexpression was observed in 85 patients (79%) with a MMR-proficient system, but only in 22 patients (48%) with a MMR-deficient colorectal cancer (P < 0.001). The lack of COX-2 overexpression was independently associated with a MMR-deficient system (odds ratio, 3.89; 95% confidence interval, 1.78-8.51; P = 0.001) and a poor degree of differentiation (OR, 3.83; 95% CI, 1.30-11.31; P = 0.015). In the subset of patients with a MMR-deficient colorectal cancer, lack of COX-2 overexpression correlated with a poor degree of differentiation, no fulfillment of Amsterdam II criteria, absence of MSH2/MLH1 germ line mutations, presence of tumor MSH2 expression, and lack of tumor MLH1 expression. CpG island promoter hypermethylation of COX2 was observed in 6 of 18 (33%) tumors lacking COX-2 expression in comparison with 2 of 28 (7%) tumors expressing this protein (P = 0.04). Up to half of MMR-deficient colorectal cancer do not show COX-2 overexpression, a fact observed almost exclusively in patients with sporadic forms. COX2 hypermethylation seems to be responsible for gene silencing in one third of them. These results suggest the potential utility of nonsteroidal anti-inflammatory drugs in HNPCC chemoprevention and may explain the lack of response of this approach in some sporadic tumors.