Merck
  • Brain-derived neurotrophic factor released from engineered mesenchymal stem cells attenuates glutamate- and hydrogen peroxide-mediated death of staurosporine-differentiated RGC-5 cells.
Brain-derived neurotrophic factor released from engineered mesenchymal stem cells attenuates glutamate- and hydrogen peroxide-mediated death of staurosporine-differentiated RGC-5 cells.
Experimental eye research (2009-06-16)
Matthew M Harper, Laura Adamson, Bas Blits, Mary Bartlett Bunge, Sinisa D Grozdanic, Donald S Sakaguchi
ABSTRACT
The purpose of this study was to determine the viability of cell-based delivery of brain-derived neurotrophic factor (BDNF) from genetically modified mesenchymal stem cells (MSCs) for neuroprotection of RGC-5 cells. RGC-5 cells were differentiated with the protein kinase inhibitor staurosporine (SS) and exposed to the cellular stressors glutamate or H2O2. As a neuroprotective strategy, these cells were then co-cultured across a membrane insert with mesenchymal stem cells (MSCs) engineered with a lentiviral vector for production of BDNF (BDNF-MSCs). As a positive control, recombinant human BDNF (rhBDNF) was added to stressed RGC-5 cells. After SS-differentiation RGC-5s developed neuronal-like morphologies, and a significant increase in the proportion of RGC-5s immunoreactive for TuJ-1 and Brn3a was observed. Differentiated RGC-5s also had prominent TrkB staining, demonstrating expression of the high-affinity BDNF receptor. Treatment of SS-differentiated RGC-5s with glutamate or H2O2, produced significant cell death (56.0 +/- 7.02 and 48.90 +/- 4.58% of control cells, respectively) compared to carrier-solution treated cells. BDNF-delivery from MSCs preserved more RGC-5 cells after treatment with glutamate (80.0 +/- 5.40% cells remaining) than control GFP expressing MSCs (GFP-MSCs, 57.29 +/- 1.89%, p < 0.01). BDNF-MSCs also protected more RGC-5s after treatment with H2O2 (65.6 +/- 3.47%) than GFP-MSCs (46.0 +/- 4.20%, p < 0.01). We have shown survival of differentiated RGC-5s is reduced by the cellular stressors glutamate and H2O2. Additionally, our results demonstrate that genetically modified BDNF-producing MSCs can enhance survival of stressed RGC-5 cells and therefore, may be effective vehicles to deliver BDNF to retinal ganglion cells affected by disease.