- A new approach to microbial production of gallic acid.
A new approach to microbial production of gallic acid.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology] (2008-10-01)
Bhakti Bajpai, Shridhar Patil
PMID24031294
ABSTRACT
In a new approach to microbial gallic acid production by Aspergillus fischeri MTCC 150, 40gL(-1) of tannic acid was added in two installments during the bioconversion phase of the process (25gL(-1) and 15gL(-1) at 32 and 44h respectively). The optimum parameters for the bioconversion phase were found to be temperature: 35°C, pH: slightly acidic (3.3-3.5), aeration: nil and agitation: 250 rpm. A maximum of 71.4% conversion was obtained after 71h fermentation with 83.3% product recovery. The yield was 7.35 g of gallic acid per g of biomass accumulated and the fermenter productivity was 0.56 g of gallic acid produced per liter of medium per hour.