Merck
  • ATM and ATR play complementary roles in the behavior of excitatory and inhibitory vesicle populations.
ATM and ATR play complementary roles in the behavior of excitatory and inhibitory vesicle populations.
Proceedings of the National Academy of Sciences of the United States of America (2017-12-28)
Aifang Cheng, Teng Zhao, Kai-Hei Tse, Hei-Man Chow, Yong Cui, Liwen Jiang, Shengwang Du, Michael M T Loy, Karl Herrup
ABSTRACT
ATM (ataxia-telangiectasia mutated) and ATR (ATM and Rad3-related) are large PI3 kinases whose human mutations result in complex syndromes that include a compromised DNA damage response (DDR) and prominent nervous system phenotypes. Both proteins are nuclear-localized in keeping with their DDR functions, yet both are also found in cytoplasm, including on neuronal synaptic vesicles. In ATM- or ATR-deficient neurons, spontaneous vesicle release is reduced, but a drop in ATM or ATR level also slows FM4-64 dye uptake. In keeping with this, both proteins bind to AP-2 complex components as well as to clathrin, suggesting roles in endocytosis and vesicle recycling. The two proteins play complementary roles in the DDR; ATM is engaged in the repair of double-strand breaks, while ATR deals mainly with single-strand damage. Unexpectedly, this complementarity extends to these proteins' synaptic function as well. Superresolution microscopy and coimmunoprecipitation reveal that ATM associates exclusively with excitatory (VGLUT1