Merck
  • NGS-based methylation profiling differentiates TCF3-HLF and TCF3-PBX1 positive B-cell acute lymphoblastic leukemia.
NGS-based methylation profiling differentiates TCF3-HLF and TCF3-PBX1 positive B-cell acute lymphoblastic leukemia.
Epigenomics (2018-01-16)
Priyadarshini Kachroo, Silke Szymczak, Femke-Anouska Heinsen, Michael Forster, Jörn Bethune, Georg Hemmrich-Stanisak, Lewis Baker, Martin Schrappe, Martin Stanulla, Andre Franke
ABSTRACT
To determine whether methylation differences between mostly fatal TCF3-HLF and curable TCF3-PBX1 pediatric acute lymphoblastic leukemia subtypes can be associated with differential gene expression and remission. Five (extremely rare) TCF3-HLF versus five (very similar) TCF3-PBX1 patients were sampled before and after remission and analyzed using reduced representation bisulfite sequencing and RNA-sequencing. We identified 7000 differentially methylated CpG sites between subtypes, of which 78% had lower methylation levels in TCF3-HLF. Gene expression was negatively correlated with CpG sites in 23 genes. KBTBD11 clearly differed in methylation and expression between subtypes and before and after remission in TCF3-HLF samples. KBTBD11 hypomethylation may be a promising potential target for further experimental validation especially for the TCF3-HLF subtype.