Merck
Prostaglandin I
Oncotarget (2017-12-22)
Shao-Qin Zheng, Zi-Yi Gong, Chen-Di Lu, Pu Wang
ABSTRACT
Cyclooxygenase-2 (COX-2) has been found to be induced during the early stage of Alzheimer's disease (AD). Using mouse-derived astrocyte and APP/PS1 transgenic (Tg) mice as model systems, we firstly elucidated the mechanisms underlying COX-2 metabolic production including prostaglandin (PG)E