- Bis(aryl) Tetrasulfides as Cathode Materials for Rechargeable Lithium Batteries.
Bis(aryl) Tetrasulfides as Cathode Materials for Rechargeable Lithium Batteries.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-09-02)
Wei Guo, Zachary D Wawrzyniakowski, Matthew M Cerda, Amruth Bhargav, Michael D Pluth, Ying Ma, Yongzhu Fu
PMID28861926
ABSTRACT
An organotetrasulfide consists of a linear chain of four sulfur atoms that could accept up to 6 e