Merck
Cobalt-catalyzed arylzincation of alkynes.
Organic letters (2009-05-12)
Kei Murakami, Hideki Yorimitsu, Koichiro Oshima
ABSTRACT
Cobalt(II) bromide catalyzes arylzincation of alkynes with arylzinc iodide x lithium chloride complexes in acetonitrile. The scope of the arylzincation is wide enough to use unfunctionalized alkynes, such as 6-dodecyne, as well as arylacetylenes. The inherent functional group compatibility of arylzinc reagents allows preparation of various functionalized styrene derivatives. The reaction is applicable to the efficient and stereoselective synthesis of a synthetic estrogen and its derivative.