- A Pd(II)-catalyzed ring-expansion reaction of cyclic 2-azidoalcohol derivatives: synthesis of azaheterocycles.
A Pd(II)-catalyzed ring-expansion reaction of cyclic 2-azidoalcohol derivatives: synthesis of azaheterocycles.
Journal of the American Chemical Society (2009-08-25)
Shunsuke Chiba, Yan-Jun Xu, Yi-Feng Wang
PMID19697931
ABSTRACT
A Pd(II)-catalyzed ring expansion-reaction of cyclic 2-azidoalcohol derivatives was found to proceed via an unprecedented C-C bond cleavage-C-N bond formation sequence, providing substituted azaheterocycles.