Merck
  • Distinct Roles of Different Presynaptic and Postsynaptic NCAM Isoforms in Early Motoneuron-Myotube Interactions Required for Functional Synapse Formation.
Distinct Roles of Different Presynaptic and Postsynaptic NCAM Isoforms in Early Motoneuron-Myotube Interactions Required for Functional Synapse Formation.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-11-28)
Katsusuke Hata, Yuka Maeno-Hikichi, Norihiro Yumoto, Steven J Burden, Lynn T Landmesser
ABSTRACT
The neural cell adhesion molecule (NCAM) is expressed both presynaptically and postsynaptically during neuromuscular junction formation. Genetic deletion in mice of all three isoforms (180, 140, and 120 kDa), or just the 180 isoform, suggested that different isoforms played distinct roles in synaptic maturation. Here we characterized in mice of either sex the earliest adhesive contacts between the growth cones of motoneurons and myotubes and their subsequent maturation into functional synapses in cocultures of motoneurons and myotubes, which expressed their normal complement of NCAM isoforms, or were lacking all isoforms either presynaptically or postsynaptically. Growth cone contact with +/+ mouse myotubes resulted in immediate adhesive contacts and the rapid downregulation of growth cone motility. When contacting NCAM