- Effects of L-Arginine Supplementation on Leukogram, Inflammatory Bowel Infiltrates and Immunoglobulins with 5-FU Use in Rats.
Effects of L-Arginine Supplementation on Leukogram, Inflammatory Bowel Infiltrates and Immunoglobulins with 5-FU Use in Rats.
Nutrition and cancer (2018-01-19)
Bianca D Balmant, Eloisa O N Araújo, Denise Yabuki, Amanda B Novais, Sandra C Genaro, Cecilia B Laposy, Paulo F I Goiozo, Marcelo G M Chacur, Rogério Giuffrida, Luis S L S Reis
PMID29345500
ABSTRACT
This study evaluated the effects of L-arginine supplementation on blood parameters, kidney and liver function, immunoglobulins and noninflammatory infiltrates in the small intestines of rats subjected to chemotherapy with 5-fluorouracil (5-FU). Thirty-two Wistar rats were randomly distributed into 4 groups (8 rats/group): an untreated control group, and test groups receiving one dose of 5-FU (G