- Palladium-catalysed dimerization of vinylarenes using indium triflate as an effective co-catalyst.
Palladium-catalysed dimerization of vinylarenes using indium triflate as an effective co-catalyst.
Chemical communications (Cambridge, England) (2003-05-13)
Teruhisa Tsuchimoto, Susumu Kamiyama, Ryoju Negoro, Eiji Shirakawa, Yusuke Kawakami
PMID12739644
ABSTRACT
A palladium-indium triflate catalyst was found to be much more active for the dimerization of vinylarenes compared with generally used cationic palladium(II) catalysts.