Merck
  • Effects of hepatitis C virus core protein and nonstructural protein 4B on the Wnt/β-catenin pathway.
Effects of hepatitis C virus core protein and nonstructural protein 4B on the Wnt/β-catenin pathway.
BMC microbiology (2017-05-27)
Xiao-Hua Jiang, Yu-Tao Xie, Ya-Ping Cai, Jing Ren, Tao Ma
ABSTRACT
Hepatitis C virus (HCV) core protein and nonstructural protein 4B (NS4B) are potentially oncogenic. Aberrant activation of the Wnt/β-catenin signaling pathway is closely associated with hepatocarcinogenesis. We investigated the effects of HCV type 1b core protein and NS4B on Wnt/β-catenin signaling in various liver cells, and explored the molecular mechanism underlying HCV-related hepatocarcinogenesis. Compared with the empty vector control, HCV core protein and NS4B demonstrated the following characteristics in the Huh7 cells: significantly enhanced β-catenin/Tcf-dependent transcriptional activity (F = 40.87, P < 0.01); increased nuclear translocation of β-catenin (F = 165.26, P < 0.01); upregulated nuclear β-catenin, cytoplasmic β-catenin, Wnt1, c-myc, and cyclin D1 protein expression (P < 0.01); and promoted proliferation of Huh7 cells (P < 0.01 or P < 0.05). Neither protein enhanced β-catenin/Tcf-dependent transcriptional activity in the LO2 cells (F = 0.65, P > 0.05), but they did significantly enhance Wnt3a-induced β-catenin/Tcf-dependent transcriptional activity (F = 64.25, P < 0.01), and promoted the nuclear translocation of β-catenin (F = 66.54, P < 0.01) and the Wnt3a-induced proliferation of LO2 cells (P < 0.01 or P < 0.05). Moreover, activation of the Wnt/β-catenin signaling pathway was greater with the core protein than with NS4B (P < 0.01 or P < 0.05). HCV core protein and NS4B directly activate the Wnt/β-catenin signaling pathway in Huh7 cells and LO2 cells induced by Wnt3a. These data suggest that HCV core protein and NS4B contribute to HCV-associated hepatocellular carcinogenesis.