- Direct carbamoylation of alkenyl halides.
Direct carbamoylation of alkenyl halides.
Organic letters (2003-12-20)
Robert F Cunico, Bikash C Maity
PMID14682736
ABSTRACT
Alkenyl chlorides and bromides are converted into tertiary enamides by treatment with a carbamoylsilane in toluene at 110 degrees C in the presence of phosphine-palladium(0) catalysts. [reaction: see text]