Merck
  • Autotransplantation of mature and immature third molars in 23 Chinese patients: a clinical and radiological follow-up study.
Autotransplantation of mature and immature third molars in 23 Chinese patients: a clinical and radiological follow-up study.
BMC oral health (2017-12-30)
Haozhe Tang, Zhengyan Shen, Minhong Hou, Ligeng Wu
ABSTRACT
We investigated the clinical and radiographic outcomes of autotransplanted teeth over a follow-up period of 2 to 8 years, and summarize the findings of other relevant studies with regard to the primary factors that influence a good prognosis in such patients. Twenty-three patients (6 men, 17 women) who attended Tanggu Stomatological Hospital, Tianjin, China, from 2008 through 2013, were included in the study. These patients presented with a variety of dental problems, including tooth loss, residual crowns, missing first or second molars, dental trauma, tooth fracture, and unrestored teeth. A total of 26 third molars, including 2 immature molars, were autotransplanted in these patients. The success rate of autotransplantation was assessed on the basis of clinical and radiographic outcomes after follow-up periods ranging from 2 to 8 years. Clinical examination revealed stability of all 26 transplanted teeth, with satisfactory masticatory function and no patient discomfort. Radiographic examination revealed normal periapical tissues and an intact lamina dura surrounding the tooth root, indicating adequate healing of periodontal tissues. Autotransplantation achieved good results in the Chinese sample population investigated, and was associated with an excellent prognosis. Rigorous case selection, adequate protection of the periodontal ligament, and proper oral hygiene contribute significantly to the long-term success of the procedure.