- Phosphorylation of the RNA-binding protein Zfs1 modulates sexual differentiation in fission yeast.
Phosphorylation of the RNA-binding protein Zfs1 modulates sexual differentiation in fission yeast.
Journal of cell science (2017-11-01)
Francisco J Navarro, Probir Chakravarty, Paul Nurse
PMID29084823
ABSTRACT
Sexual differentiation in the fission yeast