Merck
  • Preparation of Radioactive Skin Patches Using Polyhydroxamic Acid-Grafted Cellulose Films Toward Applications in Treatment of Superficial Tumors.
Preparation of Radioactive Skin Patches Using Polyhydroxamic Acid-Grafted Cellulose Films Toward Applications in Treatment of Superficial Tumors.
Cancer biotherapy & radiopharmaceuticals (2017-12-22)
Sanjay Kumar Saxena, Yogendra Kumar, Samina H Shaikh, Usha Pandey, Sanjukta A Kumar, Ashutosh Dash
ABSTRACT
The primary objective of this investigation is the development of a strategy for the synthesis of polyhydroxamic acid (PHA)-grafted cellulose film, its characterization, and evaluation of its usefulness for the preparation of