- The
The
Genes & development (2017-12-24)
Eddie Rodríguez-Carballo, Lucille Lopez-Delisle, Ye Zhan, Pierre J Fabre, Leonardo Beccari, Imane El-Idrissi, Thi Hanh Nguyen Huynh, Hakan Ozadam, Job Dekker, Denis Duboule
PMID29273679
ABSTRACT
The mammalian
The mammalian