Merck
  • High-density lipoprotein mutant eye drops for the treatment of posterior eye diseases.
High-density lipoprotein mutant eye drops for the treatment of posterior eye diseases.
Journal of controlled release : official journal of the Controlled Release Society (2017-10-11)
Kenji Suda, Tatsuya Murakami, Norimoto Gotoh, Ryosuke Fukuda, Yasuhiko Hashida, Mitsuru Hashida, Akitaka Tsujikawa, Nagahisa Yoshimura
ABSTRACT
Age-related macular degeneration (AMD), in which choroidal neovascularization (CNV) affects the center of the retina (macula), leads to the irreversible visual loss. The intravitreal injection of anti-angiogenesis antibodies improved the prognosis of AMD, but relatively less invasive therapies should be explored. In the present study, we show that a high-density lipoprotein (HDL) mutant is a therapeutically active drug carrier capable of treating a posterior eye disease in mice via instillation. Various HDL mutants were prepared with apoA-I proteins fused with different cell-penetrating peptides (CPPs) and phospholipids with different alkyl chain lengths; their sizes were further controlled in the range of 10-25nm. They were screened based on the efficiency of fluorescent dye delivery to the inner retinal layer in mice. The best mutant was found to have penetratin (PEN) as a CPP, 1,2-distearoyl-sn-glycero-3-phosphocholine (DSPC), and a size of 15nm. In preclinical studies on a laser-induced CNV murine model, 1week of instillation of the best mutant carrying the anti-angiogenesis drug pazopanib had dramatic therapeutic effects in reducing the CNV size. Importantly, the HDL mutant by itself contributed to the therapeutic effects. Future clinical trials for treating AMD with instillation of the HDL mutant are expected.