Merck
EphrinA1-Fc attenuates myocardial ischemia/reperfusion injury in mice.
PloS one (2017-12-14)
Augustin DuSablon, Justin Parks, K'Shylah Whitehurst, Heather Estes, Robert Chase, Eleftherios Vlahos, Uma Sharma, David Wert, Jitka Virag
ABSTRACT
EphrinA1, a membrane-bound receptor tyrosine kinase ligand expressed in healthy cardiomyocytes, is lost in injured cells following myocardial infarction. Previously, we have reported that a single intramyocardial injection of chimeric ephrinA1-Fc at the time of ischemia reduced injury in the nonreperfused myocardium by 50% at 4 days post-MI by reducing apoptosis and inflammatory cell infiltration. In a clinically relevant model of acute ischemia (30min)/reperfusion (24hr or 4 days) injury, we now demonstrate that ephrinA1-Fc reduces infarct size by 46% and completely preserves cardiac function (ejection fraction, fractional shortening, and chamber dimensions) in the short-term (24hrs post-MI) as well as long-term (4 days). At 24 hours post-MI, diminished serum inflammatory cell chemoattractants in ephrinA1-Fc-treated mice reduces recruitment of neutrophils and leukocytes into the myocardium. Differences in relative expression levels of EphA-Rs are described in the context of their putative role in mediating cardioprotection. Validation by Western blotting of selected targets from mass spectrometry analyses of pooled samples of left ventricular tissue homogenates from mice that underwent 30min ischemia and 24hr of reperfusion (I/R) indicates that ephrinA1-Fc administration alters several regulators of signaling pathways that attenuate apoptosis, promote autophagy, and shift from FA metabolism in favor of increased glycolysis to optimize anaerobic ATP production. Taken together, reduced injury is due a combination of adaptive metabolic reprogramming, improved cell survival, and decreased inflammatory cell recruitment, suggesting that ephrinA1-Fc enhances the capacity of the heart to withstand an ischemic insult.