Merck
  • Facile synthesis of two clay minerals supported graphitic carbon nitride composites as highly efficient visible-light-driven photocatalysts.
Facile synthesis of two clay minerals supported graphitic carbon nitride composites as highly efficient visible-light-driven photocatalysts.
Journal of colloid and interface science (2017-10-17)
Zhiming Sun, Chunquan Li, Xin Du, Shuilin Zheng, Guifang Wang
ABSTRACT
To overcome rapid electron-hole recombination and low adsorption capacity of single g-C