Merck
  • Effect of rosuvastatin on fasting and postprandial endothelial biomarker levels and microvascular reactivity in patients with type 2 diabetes and dyslipidemia: a preliminary report.
Effect of rosuvastatin on fasting and postprandial endothelial biomarker levels and microvascular reactivity in patients with type 2 diabetes and dyslipidemia: a preliminary report.
Cardiovascular diabetology (2017-11-11)
Kyoung Min Kim, Kyong Yeun Jung, Han Mi Yun, Seo Young Lee, Tae Jung Oh, Hak Chul Jang, Soo Lim
ABSTRACT
The cardiovascular benefits of statins have been proven, but their effect on circulation in small vessels has not been examined fully. We investigated the effect of 20 mg rosuvastatin on biomarkers, including paraoxonase-1 (PON-1) and asymmetric dimethylarginine (ADMA), and on microvascular reactivity. We enrolled 20 dyslipidemic patients with type 2 diabetes and 20 age- and body mass index (BMI)-matched healthy controls. Rosuvastatin (20 mg/day) was given to the patient group for 12 weeks. Biochemical parameters, including PON-1 and ADMA, were compared between the patient and control groups, and before and after rosuvastatin treatment in the patient group. Fasting and 2 h postprandial levels of PON-1 and ADMA after mixed-meal challenge were also compared. Microvascular reactivity in a peripheral artery was examined using laser Doppler flowmetry. The respective mean ± standard deviation of age and BMI were 50.1 ± 3.8 year and 25.8 ± 3.7 kg/m Rosuvastatin treatment for 12 weeks improved microvascular reactivity with concomitant beneficial changes in the postprandial levels of PON-1 and ADMA. These results suggest that rosuvastatin improves the postprandial cardiometabolic milieu in type 2 diabetes. Trial registration ClinicalTrials.gov: NCT02185963 (July 7, 2014).