Merck
Potential Causes of Elevated REE after High-Intensity Exercise.
Medicine and science in sports and exercise (2017-07-25)
Gary R Hunter, Douglas R Moellering, Stephen J Carter, Barbara A Gower, Marcas M Bamman, Lyndsey M Hornbuckle, Eric P Plaisance, Gordon Fisher
ABSTRACT
Resting energy expenditure (REE) increases after an intense exercise; however, little is known concerning mechanisms. The purpose of this study was to determine effects of a single bout of moderate-intensity continuous (MIC) aerobic exercise, or high-intensity interval (HII) exercise on REE under energy balance conditions. Thirty-three untrained premenopausal women were evaluated at baseline, after 8-16 wk of training, 22 h after either MIC (50% peak V˙O2) or HII (84% peak V˙O2). Participants were in a room calorimeter during and after the exercise challenge. Food intake was adjusted to obtain energy balance across 23 h. REE was measured after 22 h after all conditions. Twenty-three-hour urine norepinephrine concentration and serum creatine kinase activity (CrKact) were obtained. Muscle biopsies were obtained in a subset of 15 participants to examine muscle mitochondrial state 2, 3, and 4 fat oxidation. REE was increased 22 h after MIC (64 ± 119 kcal) and HII (103 ± 137 kcal). Markers of muscle damage (CrKact) increased after HII (9.6 ± 25.5 U·L) and MIC (22.2 ± 22.8 U·L), whereas sympathetic tone (urine norepinephrine) increased after HII (1.1 ± 10.6 ng·mg). Uncoupled phosphorylation (states 2 and 4) fat oxidation were related to REE (r = 0.65 and r = 0.55, respectively); however, neither state 2 nor state 4 fat oxidation increased after MIC or HII. REE was not increased after 8 wk of aerobic training when exercise was restrained for 60 h. Under energy balance conditions, REE increased 22 h after both moderate-intensity and high-intensity exercise. Exercise-induced muscle damage/repair and increased sympathetic tone may contribute to increased REE, whereas uncoupled phosphorylation does not. These results suggest that moderate- to high-intensity exercise may be valuable for increasing energy expenditure for at least 22 h after the exercise.