Merck
Eye-Drops for Activation of DREADDs.
Frontiers in neural circuits (2017-12-09)
William T Keenan, Diego C Fernandez, Lukas J Shumway, Haiqing Zhao, Samer Hattar
ABSTRACT
Designer Receptors Exclusively Activated by Designer Drugs (DREADDs) are an important tool for modulating and understanding neural circuits. Depending on the DREADD system used, DREADD-targeted neurons can be activated or repressed