Merck
C(sp
Journal of the American Chemical Society (2017-02-01)
Jennifer V Obligacion, Máté J Bezdek, Paul J Chirik
ABSTRACT
Cobalt catalysts with electronically enhanced site selectivity have been developed, as evidenced by the high ortho-to-fluorine selectivity observed in the C(sp