Merck
  • Controlled crosslinking of trimethylolpropane trimethacrylate for preparation of organic monolithic columns for capillary liquid chromatography.
Controlled crosslinking of trimethylolpropane trimethacrylate for preparation of organic monolithic columns for capillary liquid chromatography.
Electrophoresis (2017-08-24)
Mariana R Gama, Pankaj Aggarwal, Milton L Lee, Carla B G Bottoli
ABSTRACT
Organic monolithic columns based on single crosslinking of trimethylolpropane trimethacrylate (TRIM) monomer were prepared in a single step by living/controlled free-radical polymerization. Full optimization of the preparation, such as using different percentages of TRIM and different amounts of radical promoter as well as various porogen solvents were explored. The resulting monolithic columns were characterized by scanning electronic microscopy and nitrogen sorption for structure morphology studies and surface area measurements, respectively. Using capillary liquid chromatography, 150 μm i.d. columns were applied to separate a mixture of small hydrophobic molecules. The results indicated that column performance is highly sensitive to the type and the amount of porogen solvents used in the polymerization mixture composition. Good resolution factors and methylene selectivity were obtained, indicating the promising potential of this material for capillary liquid chromatography separations.