Merck
  • Injectable Polymeric Cytokine-Binding Nanowires Are Effective Tissue-Specific Immunomodulators.
Injectable Polymeric Cytokine-Binding Nanowires Are Effective Tissue-Specific Immunomodulators.
ACS nano (2017-11-11)
Colin R Zamecnik, Margaret M Lowe, David M Patterson, Michael D Rosenblum, Tejal A Desai
ABSTRACT
Injectable nanomaterials that interact with the host immune system without surgical intervention present spatially anchored complements to cell transplantation and could offer improved pharmacokinetics compared to systemic cytokine therapy. Here we demonstrate fabrication of high aspect ratio polycaprolactone nanowires coupled with cytokine-binding antibodies that assemble into porous matrices when injected into the subcutaneous space. These structures are fabricated using a nanotemplating technique that allows for tunability of particle dimensions and utilize a straightforward maleimide conjugation chemistry to allow site-specific coupling to proteins. Nanowires are well tolerated in vivo and incite minimal inflammatory infiltrate. Nanowires conjugated with antibodies were designed to capture and potentiate endogenous interleukin-2 (IL-2), an important leukocyte activating cytokine. Together these nanowire-antibody matrices were capable of localizing endogenous IL-2 in the skin and activated targeted specific natural killer and T cell subsets, demonstrating both tissue- and cell-specific immune activation. These self-assembling nanowire matrices show promise as scaffolds to present engineered, local receptor-ligand interactions for cytokine-mediated disease.