Merck
  • Etanercept for steroid-refractory acute graft-versus-host disease: A single center experience.
Etanercept for steroid-refractory acute graft-versus-host disease: A single center experience.
PloS one (2017-10-27)
Cornelis N De Jong, Lotte Saes, Clara P W Klerk, Marjolein Van der Klift, Jan J Cornelissen, Annoek E C Broers
ABSTRACT
Acute graft-versus-host disease (aGVHD) is an important complication of allogeneic stem cell transplantation (alloSCT). High dose glucocorticosteroids, are currently recommended as first-line treatment for grade II-IV aGVHD resulting in overall complete responses (CR) in 40%-50% of patients. No standard second-line regimen has been established. Different options have been reported, including anti-TNFα antibodies. We retrospectively reviewed the outcome of 15 patients with steroid-refractory (SR) aGVHD treated with etanercept at our institution. Patients were transplanted for a hematological malignancy and received either a myeloablative or a non-myeloablative conditioning regimen. Prophylaxis of GVHD consisted of cyclosporin A and mycophenolic acid. Acute GVHD was diagnosed at a median of 61 days post-transplantation. All patients had grade III aGVHD of the gut. Second-line treatment with etanercept was started at a median of 13 days after initiation of first-line therapy. Overall response rate was 53%, with CR in 3 patients and PR in 5 patients. Median overall survival after initiation of treatment with etanercept was 66 days (range 5-267) for the entire group. Median overall survival was 99 days (range 47-267 days) for responders and 17 days (range 5-66 days) for non-responders (p<0.01). Nevertheless, all patients died. Causes of death were progressive GVHD in 7 patients (47%), infection in 6 patients (40%), cardiac death in 1 patient (6.7%) and relapse in 1 patient (6,7%). Second-line treatment with etanercept does induce responses in SR-aGVHD of the gut but appears to be associated with poor long-term survival even in responding patients.