Merck
  • The butyrophilin 3A1 intracellular domain undergoes a conformational change involving the juxtamembrane region.
The butyrophilin 3A1 intracellular domain undergoes a conformational change involving the juxtamembrane region.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-07-15)
Khiem Nguyen, Jin Li, Robbins Puthenveetil, Xiaochen Lin, Michael M Poe, Chia-Hung Christine Hsiao, Olga Vinogradova, Andrew J Wiemer
ABSTRACT
Small isoprenoid diphosphates, such as (
Sigma-Aldrich
Bovine IFNG / Interferon Gamma ELISA Kit, for serum, plasma and cell culture supernatants