Merck
  • Abnormal site occupancy and high performance in warm WLEDs of a novel red phosphor, NaHF
Abnormal site occupancy and high performance in warm WLEDs of a novel red phosphor, NaHF
Dalton transactions (Cambridge, England : 2003) (2017-10-03)
Luqing Xi, Yuexiao Pan, Mengmeng Zhu, Hongzhou Lian, Jun Lin
ABSTRACT
A novel red phosphor, NaHF