- Photoluminescence, thermoluminescence glow curve and emission characteristics of Y
Photoluminescence, thermoluminescence glow curve and emission characteristics of Y
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-08-22)
Shivaramu N J, Lakshminarasappa B N, Nagabhushana K R, Swart H C, Singh Fouran
PMID28826110
ABSTRACT
Nanocrystalline Er