Merck
  • Physicochemical and adsorptive characteristics of activated carbons from waste polyester textiles utilizing MgO template method.
Physicochemical and adsorptive characteristics of activated carbons from waste polyester textiles utilizing MgO template method.
Environmental science and pollution research international (2017-08-16)
Zhihua Xu, Daofang Zhang, Zhihang Yuan, Weifang Chen, Tianqi Zhang, Danqi Tian, Haixuan Deng
ABSTRACT
Activated carbons with high specific surface areas were produced, utilizing waste polyester textiles as carbon precursor by magnesium oxide (MgO) template method. Magnesium chloride (MgCl