Merck
  • Plausible link between circa'bi'dian activity rhythms and circadian clock systems in the large black chafer
Plausible link between circa'bi'dian activity rhythms and circadian clock systems in the large black chafer
The Journal of experimental biology (2017-09-08)
Yuta Kawasaki, Hitoshi Nishimura, Sakiko Shiga
ABSTRACT
Two-day rhythms, referred to as circa'bi'dian rhythms, have been reported in humans and mosquitos. However, these rhythms only appear under constant conditions, and the functional mechanisms of 2-day rhythms were unknown. Here, we report clear circabidian rhythms of large black chafers (