- Molecular targeting of the Aurora-A/SMAD5 oncogenic axis restores chemosensitivity in human breast cancer cells.
Molecular targeting of the Aurora-A/SMAD5 oncogenic axis restores chemosensitivity in human breast cancer cells.
Oncotarget (2017-12-07)
Mateusz Opyrchal, Malgorzata Gil, Jeffrey L Salisbury, Mathew P Goetz, Vera Suman, Amy Degnim, James McCubrey, Tufia Haddad, Ianko Iankov, Chenye B Kurokawa, Nicole Shumacher, James N Ingle, Evanthia Galanis, Antonino B D'Assoro
PMID29207686
ABSTRACT
Although the majority of breast cancers initially respond to the cytotoxic effects of chemotherapeutic agents, most breast cancer patients experience tumor relapse and ultimately die because of drug resistance. Breast cancer cells undergoing epithelial to mesenchymal transition (EMT) acquire a CD44