Merck
Transforming Growth Factor-
Molecular pharmacology (2017-08-27)
Hendrik Ungefroren, David Witte, Koichiro Mihara, Bernhard H Rauch, Petra Henklein, Olaf Jöhren, Shirin Bonni, Utz Settmacher, Hendrik Lehnert, Morley D Hollenberg, Roland Kaufmann, Frank Gieseler
ABSTRACT
Transforming growth factor-