Merck
  • Environmentally friendly procedure based on VA-MSPD for the determination of booster biocides in fish tissue.
Environmentally friendly procedure based on VA-MSPD for the determination of booster biocides in fish tissue.
Food chemistry (2017-10-19)
Augusto A Vieira, Sergiane S Caldas, Ana Laura Venquiaruti Escarrone, Jean Lucas de Oliveira Arias, Ednei Gilberto Primel
ABSTRACT
Booster biocides have been widely applied to ships and other submerged structures. These compounds can be released into the marine environment as the result of vessel hull leaching and may remain in different environmental compartments. This study aimed at introducing an environmentally friendly procedure for the extraction of irgarol and diuron from fish samples by vortex-assisted matrix solid phase dispersion (VA-MSPD) with detection by liquid chromatography tandem mass spectrometry. Different types of solid supports and solvents were evaluated. The best results were found when 0.5g mussel shell, 0.5g sodium sulfate and 5mL ethanol were used. Analytical recoveries ranged from 81 to 110%, with RSD below 10%, whereas the matrix effect was between -17 and 1% (for all samples under study). LOQ values of irgarol and diuron were 5 and 50ngg