- The Potential of Alkyl Amides as Novel Biomarkers and Their Application to Paleocultural Deposits in China.
The Potential of Alkyl Amides as Novel Biomarkers and Their Application to Paleocultural Deposits in China.
Scientific reports (2017-11-09)
Jianjun Wang, Bernd R T Simoneit, Guoying Sheng, Liqi Chen, Libin Xu, Xinming Wang, Yuhong Wang, Liguang Sun
PMID29116260
ABSTRACT
A series of alkyl amides was detected and identified in the sedimentary record from an archaeological site at Yuchisi, Mengcheng, Anhui, China. The alkyl amides profiles change abruptly at the depth corresponding to the transition between two prehistoric cultures, which also corresponds to an abrupt change in the fatty acid ratio C