- Checkpoint kinase 2 is dispensable for regulation of the p53 response but is required for G
Checkpoint kinase 2 is dispensable for regulation of the p53 response but is required for G
Apoptosis : an international journal on programmed cell death (2017-07-25)
Yukihiro Furusawa, Yuka Yamanouchi, Takashi Iizumi, Qing-Li Zhao, Yohei Mitsuhashi, Akinori Morita, Atushi Enomoto, Yoshiaki Tabuchi, Takashi Kondo
PMID28733865
ABSTRACT
Hyperthermia induced by heat stress (HS) is known to inhibit proliferation and induce cell death in cancer. We previously demonstrated that checkpoint kinase 1 (Chk1) contributes to G