Merck
  • Decreased Prostaglandin D2 Levels in Major Depressive Disorder Are Associated with Depression-Like Behaviors.
Decreased Prostaglandin D2 Levels in Major Depressive Disorder Are Associated with Depression-Like Behaviors.
The international journal of neuropsychopharmacology (2017-06-06)
Cuilin Chu, Hui Wei, Wanwan Zhu, Yan Shen, Qi Xu
ABSTRACT
Prostaglandin (PG) D2 is the most abundant prostaglandin in the mammalian brain. The physiological and pharmacological actions of PGD2 in the central nervous system seem to be associated with some of the symptoms exhibited by patients with major depressive disorder. Previous studies have found that PGD2 synthase was decreased in the cerebrospinal fluid of major depressive disorder patients. We speculated that there may be a dysregulation of PGD2 levels in major depressive disorder. Ultra-performance liquid chromatography-tandem mass spectrometry coupled with a stable isotopic-labeled internal standard was used to determine PGD2 levels in the plasma of major depressive disorder patients and in the brains of depressive mice. A total of 32 drug-free major depressive disorder patients and 30 healthy controls were recruited. An animal model of depression was constructed by exposing mice to 5 weeks of chronic unpredictable mild stress. To explore the role of PGD2 in major depressive disorder, selenium tetrachloride was administered to simulate the change in PGD2 levels in mice. Mice exposed to chronic unpredictable mild stress exhibited depression-like behaviors, as indicated by reduced sucrose preference and increased immobility time in the forced swimming test. PGD2 levels in the plasma of major depressive disorder patients and in the brains of depressive mice were both decreased compared with their corresponding controls. Further inhibiting PGD2 production in mice resulted in an increased immobility time in the forced swimming test that could be reversed by imipramine. Decreased PGD2 levels in major depressive disorder are associated with depression-like behaviors.