- Progesterone receptor membrane component 1 is phosphorylated upon progestin treatment in breast cancer cells.
Progesterone receptor membrane component 1 is phosphorylated upon progestin treatment in breast cancer cells.
Oncotarget (2017-10-27)
Marina Willibald, Giuliano Bayer, Vanessa Stahlhut, Gereon Poschmann, Kai Stühler, Berthold Gierke, Michael Pawlak, Harald Seeger, Alfred O Mueck, Dieter Niederacher, Tanja Fehm, Hans Neubauer
PMID29069804
ABSTRACT
Menopausal hormone therapy, using estrogen and synthetic progestins, is associated with an increased risk of developing breast cancer. The effect of progestins on breast cells is complex and not yet fully understood. In previous