Merck
Ionic effects on the proton transfer mechanism in aqueous solutions.
Physical chemistry chemical physics : PCCP (2017-09-14)
Joonyoung F Joung, Sangin Kim, Sungnam Park
ABSTRACT
Proton dissociation (PD) reactions of weak acids and proton transfer (PT) processes in aqueous solutions are strongly influenced by ions. However, a detailed molecular picture that describes how ions affect the rates of PD and PT processes is still missing. Here, we utilize time-resolved fluorescence spectroscopy combined with quantum chemical calculations to investigate the excited-state proton transfer (ESPT) reaction of a photoacid in aqueous metal chloride solutions. The activation energy (E