Merck
  • Cap-n-Collar Promotes Tissue Regeneration by Regulating ROS and JNK Signaling in the
Cap-n-Collar Promotes Tissue Regeneration by Regulating ROS and JNK Signaling in the
Genetics (2017-05-18)
Amanda R Brock, Mabel Seto, Rachel K Smith-Bolton
ABSTRACT
Regeneration is a complex process that requires an organism to recognize and repair tissue damage, as well as grow and pattern new tissue. Here, we describe a genetic screen to identify novel regulators of regeneration. We ablated the