Merck
  • Imaging thiol redox status in murine tumors in vivo with rapid-scan electron paramagnetic resonance.
Imaging thiol redox status in murine tumors in vivo with rapid-scan electron paramagnetic resonance.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2017-01-17)
Boris Epel, Subramanian V Sundramoorthy, Martyna Krzykawska-Serda, Matthew C Maggio, Mark Tseytlin, Gareth R Eaton, Sandra S Eaton, Gerald M Rosen, Joseph P Y Kao, Howard J Halpern
ABSTRACT
Thiol redox status is an important physiologic parameter that affects the success or failure of cancer treatment. Rapid scan electron paramagnetic resonance (RS EPR) is a novel technique that has shown higher signal-to-noise ratio than conventional continuous-wave EPR in in vitro studies. Here we used RS EPR to acquire rapid three-dimensional images of the thiol redox status of tumors in living mice. This work presents, for the first time, in vivo RS EPR images of the kinetics of the reaction of