Merck
  • Cathepsin K knockout protects against cardiac dysfunction in diabetic mice.
Cathepsin K knockout protects against cardiac dysfunction in diabetic mice.
Scientific reports (2017-08-20)
Rui Guo, Yinan Hua, Olivia Rogers, Travis E Brown, Jun Ren, Sreejayan Nair
ABSTRACT
Diabetes is a major risk factor for cardiovascular disease and the lysosomal cysteine protease cathepsin K plays a critical role in cardiac pathophysiology. To expand upon our previous findings, we tested the hypothesis that, knockout of cathepsin K protects against diabetes-associated cardiac anomalies. Wild-type and cathepsin K knockout mice were rendered diabetic by streptozotocin (STZ) injections. Body weight, organ mass, fasting blood glucose, energy expenditure, cardiac geometry and function, cardiac histomorphology, glutathione levels and protein levels of cathepsin K and those associated with Ca