- Dovitinib enhances temozolomide efficacy in glioblastoma cells.
Dovitinib enhances temozolomide efficacy in glioblastoma cells.
Molecular oncology (2017-05-14)
Thatchawan Thanasupawat, Suchitra Natarajan, Amy Rommel, Aleksandra Glogowska, Hugo Bergen, Jerry Krcek, Marshall Pitz, Jason Beiko, Sherry Krawitz, Inder M Verma, Saeid Ghavami, Thomas Klonisch, Sabine Hombach-Klonisch
PMID28500786
ABSTRACT
The multikinase inhibitor and FDA-approved drug dovitinib (Dov) crosses the blood-brain barrier and was recently used as single drug application in clinical trials for GB patients with recurrent disease. The Dov-mediated molecular mechanisms in GB cells are unknown. We used GB patient cells and cell lines to show that Dov downregulated the stem cell protein Lin28 and its target high-mobility group protein A2 (HMGA2). The Dov-induced reduction in pSTAT3