- Extreme polarisation sensitivity in the retina of the corn borer moth
Extreme polarisation sensitivity in the retina of the corn borer moth
The Journal of experimental biology (2017-03-28)
Gregor Belušič, Katja Šporar, Andrej Meglič
PMID28341662
ABSTRACT
The visual system of the European corn borer (